Learn More
Chemglass Inc OptiChem Heating Mantle Controller, Single Circuit, 30/60VAC Variable Output Voltage, 1A, 60 Watts, 120VAC Supply Voltage
Numéro de catalogue. 50401789
Encompass
OptiChem Heating Mantle Controller, Single Circuit, 30/60VAC Variable Output Voltage, 1A, 60 Watts, 120VAC Supply Voltage. Similar to CG-15005-01, but the voltage output can be adjusted between 30 and 60V. Controller is 4 W x 5 D x 3.7 H. All units are supplied with a 6ft separable power cord, a grounded, 3-wire receptacle and lighted power switch. Controller for use with CG-14000 and CG-14002 Heating Mantles rated at 30 or 60 Volts. 3-Year warranty.OptiChem is a registered Trademark of Chemglass, Inc.
Numéro de catalogue. 50401789 Fournisseur Chemglass Inc Code fournisseur CG1500620
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
OptiChem Heating Mantle Controller, Single Circuit, 30/60VAC Variable Output Voltage, 1A, 60 Watts, 120VAC Supply Voltage. Similar to CG-15005-01, but the voltage output can be adjusted between 30 and 60V. Controller is 4 W x 5 D x 3.7 H. All units are supplied with a 6ft separable power cord, a grounded, 3-wire receptacle and lighted power switch. Controller for use with CG-14000 and CG-14002 Heating Mantles rated at 30 or 60 Volts. 3-Year warranty.OptiChem is a registered Trademark of Chemglass, Inc.