missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Optical Heat Seal, for Piko PCR plates

Catalog No. HSF0031
Change view
Click to view available options
Quantity:
100 sheets
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
HSF0031 100 sheets
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. HSF0031 Supplier Thermo Scientific™ Supplier No. HSF0031

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The exceptionally clear Thermo Scientific Piko PCR/qPCR Optical Heat Seals allow high transmission of fluorescence across the range of commonly-used fluorophores

When used with a Piko Thermal Cycler or PikoReal Real-Time PCR System, sample volumes as low as 5 μL can be used with minimal evaporative loss. These optical films are available in the ¼-size format for Piko PCR Plates.

Features of the Thermo Scientific Piko PCR/qPCR Film Seals
• Optical clarity with minimal auto-fluorescence for real-time PCR applications
• Tight sealing for sample volumes as low as 5 μL
• Cryo-compatible adhesive is effective to –80°C

Related Products
Optical Adhesive Film, for Piko PCR plates

Specifications

Color Optical
Quantity 100 sheets
For Use With (Equipment) Piko Thermal Cycler or PikoReal Real-Time PCR System
Product Type Film, Seal, or Applicator
Product Line Piko
Adhesive Optical Heat Seal
Temperature Range (Metric) -80°C
Plate Compatibility Piko Plate