missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Optical Glass Lens Set with Storage Box, Set of 6

Optical glass lens set with storage box includes 6 different optical glass lenses.

Supplier:  Eisco™ PH0538B

 View more versions of this product

Catalog No. S27408


Only null left
Explore available promotions

Description

Description

  • Set includes one of each: double convex, plano-convex, converging concave-convex, diverging concave-convex, double concave, plano-concave
  • Ground and polished face, ground edges
  • Includes wooden storage case with latch
Specifications

Specifications

Glass
1.5 in.
Glass Lens Set
38 mm
Wooden Storage Box, and One of Each Lens Type: Double Convex, Plano-Convex, Converging Concave-Convex, Diverging Concave-Convex, Double Concave, Plano-Concave
Spherical
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions