Learn More
Thermo Scientific™ Optical Adhesive Film, for Piko PCR plates
$533.51
Specifications
Color Optical
Product Type Film, Seal, or Applicator
Product Line Piko
Adhesive Pressure Sensitive Optical Seal
Plate Compatibility Piko Plate
View More Specs
Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
ASF0020
View Documents Promotion Details
Thermo Scientific™
ASF0020
400 Sheets
Case of 400 Each for $533.51
Only null left
Description
Description
The exceptionally clear Thermo ScientificPiko PCR/qPCR Film Seal allows high transmission of fluorescence across the range of commonly-used fluorophores. When used with aPiko Thermal CyclerorPikoReal Real-Time PCR System, sample volumes as low as 5μL can be used with minimal evaporative loss. These optical adhesive films are available in the ¼-size format for Piko PCR Plates.
Highlights
  • Optical clarity with minimal auto-fluorescence for real-time PCR applications
  • Tight sealing for sample volumes as low as 5μL
  • Adhesive backing allows easy application without costly sealing devices
  • Cryo-compatible adhesive is effective to -80°C
Specifications
Specifications
Optical
Piko
Piko Plate
Film, Seal, or Applicator
Pressure Sensitive Optical Seal