missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eisco™ Optical Acrylic Lens Set with Storage Box, Set of 6
Optical acrylic lens set with storage box includes 6 different optical acrylic lenses.
Supplier: Eisco™ PH0538D
Description
- Set includes one of each: double convex, plano-convex, converging concave-convex, diverging concave-convex, double concave, plano-concave
- Polished face and edges
- Includes wooden storage case with latch
Specifications
| Acrylic | |
| 1.5 in. | |
| Optical Lens Set |
| 38 mm | |
| Wooden Storage Box, and One of Each Lens Type: Double Convex, Plano-Convex, Converging Concave-Convex, Diverging Concave-Convex, Double Concave, Plano-Concave | |
| Spherical |