missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Opti-MEM™ I Reduced Serum Medium, GlutaMAX™ Supplement
GREENER_CHOICE

Opti-MEM™ I Reduced-Serum Medium is an improved Minimal Essential Medium (MEM) that allows for a reduction of Fetal Bovine Serum supplementation by at least 50% with no change in growth rate or morphology.

$109.00 - $1,566.00

Specifications

Cell Line Sp2, AE-1, CHO, BHK-21, and HEK
Cell Type Primary Fibroblasts
Classification Animal Origin
Concentration 1X
Form Liquid
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
51985034
GREENER_CHOICE
Encompass
View Documents Promotion Details
Gibco™
51985034
500 mL
Each for $109.00
Only null left
 
51985091
GREENER_CHOICE
Encompass
View Documents Promotion Details
Gibco™
51985091
10 x 500 mL
Each for $1,566.00
Only null left
 
Description

Description

Opti-MEM™ I Reduced-Serum Medium is an improved Minimal Essential Medium (MEM) that allows for a reduction of Fetal Bovine Serum supplementation by at least 50% with no change in growth rate or morphology. Opti-MEM™ I medium is also recommended for use with cationic lipid transfection reagents, such as Lipofectamine™ reagent. Opti-MEM™ I medium can be used with a variety of suspension and adherent mammalian cells, including Sp2, AE-1, CHO, BHK-21, HEK, and primary fibroblasts. We offer a variety of Opti-MEM™ I modifications for a range of cell culture applications.

This Opti-MEM™ I is modified as follows:.

With: GlutaMAX™, Phenol Red.

The complete formulation is confidential. For more information, please contact Technical Services.

Using Opti-MEM™ I Medium.

Opti-MEM™ I Reduced-Serum Medium is a unique medium that contains insulin, transferrin, hypoxanthine, thymidine, and trace elements. These additional components allow for a reduction in serum supplementation by at least 50%. Opti-MEM™ I medium uses a sodium bicarbonate buffer system (2.4 g/L), and therefore requires a 5–10% CO2 environment to maintain physiological pH.

Specifications

Specifications

Sp2, AE-1, CHO, BHK-21, and HEK
Animal Origin
Liquid
Sterile-filtered
cGMP-compliant under the ISO 13485 standard
Room Temperature
Primary Fibroblasts
1X
Opti-MEM I
GlutaMAX, Phenol Red
Gibco, GlutaMAX, Opti-MEM
Storage conditions: 2-8° C. Protect from light
Shipping conditions: Ambient
Shelf life: 18 months from date of manufacture
Videos
Safety and Handling

Safety and Handling

ShelfLife : 18 Months From Date of Manufacture

SDS
Documents

Documents

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.