Learn More
Justrite™ Polypropylene, Platinum-cured Silicone, Open Top Carboy Cap with Two 1/8 in. Molded-In Hose Barbs and Tubing for Aspetic Fluid Handling, 53 mm
GREENER_CHOICE
Catalog No. 19830484
Change view
Click to view available options
Material:
Polypropylene, Platinum-cured Silicone
Catalog No. Material
19830484 Polypropylene, Platinum-cured Silicone
1 options
Catalog No. 19830484 Supplier Justrite™ Supplier No. JMC12885
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
For Use With: Compatible on bottles and carboys with 53-mm, 83-mm, or 120-mm closures
  • Helps avoid open-air pouring with leak-proof closed system
  • Pre-mounted platinum-cured silicone tubing for easy custimization of carboys
Specifications
Description Open top cap with adapter, Two 1/8 in. molded-in hose barbs and 12 in. tubing
Diameter (English) Outer 3 in.
For Use With (Equipment) Compatible on bottles and carboys with 53-mm, 83-mm, or 120-mm closures
Height (English) 13.1 in.
Material Polypropylene, Platinum-cured Silicone
Product Type Cap with Tubing