missing translation for 'onlineSavingsMsg'
Learn More

Justrite™ Polypropylene, Platinum-cured Silicone, Open Top Carboy Cap with Two 1/4 in. Molded-In Hose Barbs and Tubing for Aspetic Fluid Handling, 120 mm
GREENER_CHOICE

Catalog No. 19830487
Change view
Click to view available options
Diameter (English) Outer:
7.5 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (English) Outer
19830487 7.5 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19830487 Supplier Justrite™ Supplier No. JMC12889

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For Use With: Compatible on bottles and carboys with 53-mm, 83-mm, or 120-mm closures

  • Helps avoid open-air pouring with leak-proof closed system
  • Pre-mounted platinum-cured silicone tubing for easy custimization of carboys

Specifications

Description Open top cap with adapter, Two 1/4 in. molded-in hose barbs and 37 in. tubing
Diameter (English) Outer 7.5 in.
For Use With (Equipment) Compatible on bottles and carboys with 53-mm, 83-mm, or 120-mm closures
Height (English) 38.1 in.
Material Polypropylene, Platinum-cured Silicone
Product Type Cap