missing translation for 'onlineSavingsMsg'
Learn More

PIG™ Open-Head UN Rated Steel Pail

Catalog No. 18808758
Change view
Click to view available options
Capacity (English):
5 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English)
18808758 5 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18808758 Supplier PIG™ Supplier No. DRM1064

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A durable option for storing solids, this steel pail features a quick-release lever lock.

  • 24-gauge steel provides reliable strength
  • UN rated for Packing Groups II and III; ideal for storing and shipping hazardous or non-hazardous solids
  • Steel fold-down handle makes carrying easy
  • Quick-release lever lock band can be wire sealed to deter unauthorized access

Specifications

Product Type Open Head Pail
Capacity (English) 5 gal.
Capacity (Metric) 19 L
Color Gray
Material Steel