missing translation for 'onlineSavingsMsg'
Learn More

Youngstown Barrel & Drum Open Head Steel Drum

Catalog No. 18931A
Change view
Click to view available options
Capacity (Metric):
18.92 L
114 L
208.19 L
321.76 L
Color:
Black
Black, White
Gray
Yellow
Material:
Carbon Steel
Steel
Certifications/Compliance:
UN 1A2/X428/S-DOT/UN
UN 1A2/Y1.5/150/X250/S-DOT/UN
UN 1A2/Y1.6/150/X400/S-DOT/UN
UN 1A2/Y27/S-DOT/UN
UN A2/Y1.5/150, UN 1A2/X400/S-DOT/UN
UN A2/Y1.6/150, UN 1A2/x400/S-DOT/UN
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Color Material Certifications/Compliance
18931A 18.92 L Gray Steel UN 1A2/Y27/S-DOT/UN
191304692 208.19 L Black, White Steel UN 1A2/Y1.6/150/X400/S-DOT/UN
18930C 114 L Black Carbon Steel UN A2/Y1.5/150, UN 1A2/X400/S-DOT/UN
191304693 321.76 L Yellow Steel UN 1A2/X428/S-DOT/UN
191304849 208.19 L Black Steel UN 1A2/Y1.6/150/X400/S-DOT/UN
18929B 208.19 L Black Carbon Steel UN A2/Y1.6/150, UN 1A2/x400/S-DOT/UN
191304689 114 L Black, White Steel UN 1A2/Y1.5/150/X250/S-DOT/UN
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 18931A Supplier Youngstown Barrel & Drum Supplier No. 2169

Youngstown Open Head Steel Drum is designed for the transport of hazardous solids or liquids.

Youngstown Open Head Steel Drum is designed for the transport of hazardous solids or liquids.

With cold-rolled carbon-steel construction, steel drums are qualified for safe transport of noncorrosive hazardous liquids or solids.

  • Salvage drum designed to overpack and ship damaged and/or leaking drums
  • Also qualify for direct containment of hazardous solids
  • Freight Class 50

Specifications

Certifications/Compliance UN 1A2/Y27/S-DOT/UN
Product Type Pail
Closure Type Side Lever Lock Ring
Description 26 Gauge
Capacity (Metric) 18.92 L
Color Gray
For Use With (Application) For transport of hazardous solids or liquids, Designed to overpack and ship damaged and/or leaking drums
Material Steel
Freight Class 50
Head Style Open
Quantity Ea.
Height (English) 12.87 in.
Height (Metric) 32.7 cm
Diameter (English) Inner 11.25 in.
Diameter (Metric) Inner 28.57 cm
Weight (English) 5 lb.
Weight (Metric) 2.26 Kg
Show More Show Less