missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Youngstown Barrel & Drum Open Head Steel Drum
Youngstown Open Head Steel Drum is designed for the transport of hazardous solids or liquids.
Supplier: Youngstown Barrel & Drum YBD445
Description
Youngstown Open Head Steel Drum is designed for the transport of hazardous solids or liquids.
With cold-rolled carbon-steel construction, steel drums are qualified for safe transport of noncorrosive hazardous liquids or solids.
- Salvage drum designed to overpack and ship damaged and/or leaking drums
- Also qualify for direct containment of hazardous solids
- Freight Class 50
Chemical Identifiers
| For transport of hazardous solids or liquids, Designed to overpack and ship damaged and/or leaking drums | |
| 50 | |
| Open | |
| Ea. |
Specifications
| UN A2/Y1.5/150, UN 1A2/X400/S-DOT/UN | |
| Bolt Ring | |
| 114 L | |
| For transport of hazardous solids or liquids, Designed to overpack and ship damaged and/or leaking drums | |
| 50 | |
| Ea. | |
| 71.12 cm | |
| 46.35 cm | |
| 15.87 Kg |
| Unlined Drum | |
| 18 Gauge | |
| Black | |
| Carbon Steel | |
| Open | |
| 28 in. | |
| 18.25 in. | |
| 35 lb. |