missing translation for 'onlineSavingsMsg'
Learn More

Dunlop™ Onguard™ Wader Work Boots

Catalog No. 19024380
Change view
Click to view available options
Size:
10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19024380 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19024380 Supplier Dunlop™ Supplier No. 860561000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Flexible in cold weather

  • Waterproof hip waders with built-in steel shanks
  • Durable PVC uppers with polyester backing
  • Can be folded or rolled with no cracking
  • Resist oil, grease, gasoline, and salt water
  • Nonconductive adjustable polyester-nylon strap with plastic snaps
  • Cleated soles
  • Black

Compliance

Steel toe meets or exceeds ASTM™ F2413-11

Specifications

Certifications/Compliance ASTM F2413-11
Color Black
For Use With (Application) For utilities, municipalities, aquaculture and commercial fishing
Includes Plastic snaps, non-absorbent lining
Material PVC, Polyester
Seam Type Seamless
Shank Material Steel
Size 10
Sole Features Cleated outsole, Steel midsole, replaceable cushion insole
Strap Material Polyester Nylon
Toe Material Steel
Resistant To Oil, grease, gasoline, salt water
Show More Show Less