Learn More
Dunlop™ Onguard™ PVC Boot Covers
Catalog No. 17986302
Change view
Click to view available options
Size:
Large
X-Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17986302 X-Large
17986301 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17986302 Supplier Dunlop™ Supplier No. 97590XL00
Only null left
Waterproof boot covers made of 0.2 mm thick PVC plastic uppers heat welded to 0.5 mm thick embossed PVC soles.
  • Waterproof
  • 15 in. high (38cm) boots with elastic top
  • Yellow
Specifications
Size X-Large
Material PVC
Color Yellow
Disposable Reusable
Sterility Non-sterile
Closure Type Elastic Top
Sole Type PVC
Packaging Quantity 1 Pair/Pk.
Height (English) 15 in.
Height (Metric) 38.1 cm
Resistance Water
Product Type Boot Covers
Show More Show Less