missing translation for 'onlineSavingsMsg'
Learn More

Dunlop™ Onguard™ DuraPro™ XCP Chemical Resistant Steel Toe Boots with Steel Shank

Catalog No. p-5234110
Change view
Click to view available options
Size:
10
12
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
18500549 10
18500551 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18500549 Supplier Dunlop™ Supplier No. 8408610
Only null left

High performance compund provides ideal chemical resistance in the upper and superior abrasion and slip resistance in the outsole

  • Sloped comfort top rim prevents chafing and reduces discomfort when walking, bending and kneeling
  • 360degree pull tab for easy donning
  • Steel shank protects and supports arch
  • Larger top circumference for quick and easy pull-on
  • 3/4 in. height inner liner allows  increased flexibility, subtle feel at the calf, and soft lined feel of leg and foot
  • Honeycomb design molded inside the heel to reduce impact when walking
  • Ergonomically designed foot-bed to provide better underfoot support
  • Protective bumpers at rear of boot to prevent premature wear/puncture
  • Extended circumference kick-off ledge
  • Composition: Superpoly Upper; Sureflex Outsole
  • Sole Design: Next Generation Chevron
  • Available in sizes ranging from 3 to 16

Specifications

Certifications/Compliance F2413-11 Compliant
Color Brown
Material Superpoly, Sureflex
Shank Material Steel
Size 10
Sole Features Next Generation Chevron, slip-resistant
Toe Style Steel
Resistant To Chemicals
Product Type Chemical Resistant Footwear