Learn More
Dunlop™ Onguard™ DuraPro™ Chemical Resistant Steel Toe Boots with Steel Shank
Description
- Sloped comfort top rim prevents chafing and reduces discomfort when walking, bending and kneeling
- 360-degree pull tab for easy donning
- Larger top circumference for quick and easy pull-on
- 3/4 in. height inner liner allows increased flexibility, subtle feel at the calf, and soft lined feel of leg and foot
- Honeycomb design molded inside the heel to reduce impact when walking
- Polyblend Upper; Sureflex Outsole
- Ergonomically designed foot-bed to provide better underfoot support
- Protective bumpers at rear of boot to prevent premature wear/puncture
- Extended circumference kick-off ledge
Specifications
Specifications
| Color | Blue |
| Material | Polyblend, Sureflex |
| Shank Material | Steel shank |
| Size | 9 |
| Sole Features | Next Generation Safety-loc |
| Toe Style | Steel toe |
| Resistant To | Chemicals |