Learn More
Dunlop™ Onguard™ DuraPro™ Chemical Resistant Steel Toe Boots with Steel Shank
Catalog No. 18500515
Change view
Click to view available options
Size:
10
11
12
13
16
4
7
8
9
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
18500515 4
18500518 7
18500519 8
18500520 9
18500521 10
18500522 11
18500523 12
18500524 13
18500527 16
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 18500515 Supplier Dunlop™ Supplier No. 890864
High performance steel toe boot provides ideal chemical resistance in the upper and superior abrasion and slip resistance in the outsole
  • Sloped comfort top rim prevents chafing and reduces discomfort when walking, bending and kneeling
  • 360-degree pull tab for easy donning
  • Larger top circumference for quick and easy pull-on
  • 3/4 in. height inner liner allows  increased flexibility, subtle feel at the calf, and soft lined feel of leg and foot
  • Honeycomb design molded inside the heel to reduce impact when walking
  • Polyblend Upper; Sureflex Outsole
  • Ergonomically designed foot-bed to provide better underfoot support
  • Protective bumpers at rear of boot to prevent premature wear/puncture
  • Extended circumference kick-off ledge
Specifications
Color Blue
Material Polyblend, Sureflex
Shank Material Steel shank
Size 4
Sole Features Next Generation Safety-loc
Toe Style Steel
Resistant To Chemicals