missing translation for 'onlineSavingsMsg'
Learn More

Dunlop™ Onguard™ Hazmax™ Boots

Catalog No. p-38272
Change view
Click to view available options
Color:
Yellow
Size:
10
12
7
Height (English):
16 in.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size Height (English)
17985403C Yellow 7 16 in.
17985403F Yellow 10 16 in.
17985403H Yellow 12 16 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Do not use for fire protection.
Catalog No. 17985403C Supplier Dunlop™ Supplier No. 870120700
Only null left
Do not use for fire protection.

Choose from two styles

Ideal for emergency response teams, hazmat technicians, environmental specialists, and site remediation workers.

Over-the-sock knee boots

  • Steel toes
  • Full steel midsoles and steel ladder shanks
  • Limited flash fire protection; for escape only in the event of a chemical fire
  • Preformed three-layer comfort insoles
  • Kick-off lug
  • Green chemical-resistant uppers
  • Yellow Ultragrip™ outsoles
  • No snaps or buttons for more reliable fit
Overboot
  • Adjustable nonconductive polyester straps
  • Nonmetallic buckles and snaps
  • Kick-off lug
  • Black cleated outsoles

Order Info

When ordering over-the-sock boots to be worn over a chemical protective suit with attached booties, order two shoe sizes larger. When ordering overboots, order regular shoe size. Results of chemical permeation tests available by calling Fisher Safety Customer Service at 1-800-772-6733.

Compliance

Meets ASTM™ F2413-05; CSA Z195 requirements.

Specifications

Color Yellow
For Use With (Application) Ideal for emergency response teams, hazmat technicians, environmental specialists, and site remediation workers.
Height (English) 16 in.
Material Flexible PVC Alloy
Size 7