Learn More
New England Biolab Ltd OneTaq RT-PCR Kit 30 reactions
Numéro de catalogue. 50591131
Encompass
OneTaq RT-PCR Kit combines two powerful mixes, M-MuLV Enzyme Mix and OneTaq Hot Start 2X Master Mix with Standard Buffer for 2-step RT-PCR applications. The two mixes require minimal handling during reaction setup and yet offer consistent and robust RT-PCR reactions. The first strand cDNA synthesis is achieved by using two optimized mixes, M-MuLV Enzyme Mix and M-MuLV Reaction Mix. M-MuLV Enzyme Mix combines M-MuLV Reverse Transcriptase and murine RNase Inhibitor while M-MuLV Reaction Mix contains dNTPs and an optimized buffer. The kit also contains two optimized primers for reverse transcription and nuclease-free water. An anchored oligo-dT primer [d(T)23VN] forces the primer to anneal to the beginning of the polyA tail. The optimized Random Primer Mix provides random and consistent priming sites covering the entire RNA templates including both mRNAs and non-polyadenylated RNAs. The amplification step features a OneTaq Hot Start DNA Polymerase in a master mix format.
Numéro de catalogue. 50591131 Fournisseur New England Biolab Ltd Code fournisseur E5310S
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
OneTaq RT-PCR Kit combines two powerful mixes, M-MuLV Enzyme Mix and OneTaq Hot Start 2X Master Mix with Standard Buffer for 2-step RT-PCR applications. The two mixes require minimal handling during reaction setup and yet offer consistent and robust RT-PCR reactions. The first strand cDNA synthesis is achieved by using two optimized mixes, M-MuLV Enzyme Mix and M-MuLV Reaction Mix. M-MuLV Enzyme Mix combines M-MuLV Reverse Transcriptase and murine RNase Inhibitor while M-MuLV Reaction Mix contains dNTPs and an optimized buffer. The kit also contains two optimized primers for reverse transcription and nuclease-free water. An anchored oligo-dT primer [d(T)23VN] forces the primer to anneal to the beginning of the polyA tail. The optimized Random Primer Mix provides random and consistent priming sites covering the entire RNA templates including both mRNAs and non-polyadenylated RNAs. The amplification step features a OneTaq Hot Start DNA Polymerase in a master mix format.