Learn More
New England Biolab Ltd OneTaq Hot Start 2X Master Mix with Standard Buffer 100 reactions (50 l vol)
Catalog No. NC0108401
Encompass
OneTaq Hot Start 2X Master Mix with Standard Buffer is an optimized blend of Taq and Deep Vent DNA Polymerases combined with an aptamer-based inhibitor. This enzyme blend is ideally suited to routine PCR applications from a variety of templates, including pure DNA solutions, bacterial colonies and cDNA products. The 3' to 5' exonuclease activity of Deep Vent DNA Polymerase increases the fidelity and robust amplification of Taq DNA Polymerase. The hot start nature of the enzyme offers convenience with decreased interference from primer dimers and secondary products. The convenient master mix formulation contains dNTPs, MgCl2 and other buffer components and stabilizers listed below, requiring only the addition of primers and DNA template for robust amplification.
Catalog No. NC0108401 Supplier New England Biolab Ltd Supplier No. M0484S
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
OneTaq Hot Start 2X Master Mix with Standard Buffer is an optimized blend of Taq and Deep Vent DNA Polymerases combined with an aptamer-based inhibitor. This enzyme blend is ideally suited to routine PCR applications from a variety of templates, including pure DNA solutions, bacterial colonies and cDNA products. The 3' to 5' exonuclease activity of Deep Vent DNA Polymerase increases the fidelity and robust amplification of Taq DNA Polymerase. The hot start nature of the enzyme offers convenience with decreased interference from primer dimers and secondary products. The convenient master mix formulation contains dNTPs, MgCl2 and other buffer components and stabilizers listed below, requiring only the addition of primers and DNA template for robust amplification.