missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ondansetron Related Compound A, Certified Reference Material, MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 119812-29-2 |
| Flash Point | Not Applicable |
| Molecular Formula | C16H20N2O · HCl |
| Linear Formula | C16H20N2O · HCl |
| Quantity | 100 mg |
| MDL Number | MFCD00800231 |
| Synonym | 3-[(Dimethylamino)methyl]-9-methyl-1,2,3,9-tetrahydro-4 H-carbazol-4-one hydrochloride; 3-[(Dimethylamino)methyl]-1,2,3,9-tetrahydro-9-methyl-4 H-carbazol-4-one |
| Formula Weight | 292.80 |
| Grade | Certified Reference Material |