missing translation for 'onlineSavingsMsg'
Learn More

Ondansetron Hydrochloride Dihydrate 98.0+%, TCI America™

Catalog No. O04071G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O04071G 1 g
O04075G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O04071G Supplier TCI America Supplier No. O04071G

Chemical Identifiers

CAS 103639-04-9
Molecular Formula C18H23ClN3O3
Molecular Weight (g/mol) 364.85
MDL Number MFCD00374371
InChI Key BGGIFKYQGVGSIW-UHFFFAOYSA-N
Synonym 1,2,3,9-tetrahydro-9-methyl-3-2-methyl-1h-imidazol-1-yl methyl-4h-carbazol-4-one hydrochloride; gr 38032f; ondansetron hydrochloride
PubChem CID 71317260
IUPAC Name chlorine;9-methyl-3-[(2-methylimidazol-1-yl)methyl]-2,3-dihydro-1H-carbazol-4-one;dihydrate
SMILES CC1=NC=CN1CC2CCC3=C(C2=O)C4=CC=CC=C4N3C.O.O.[Cl]

Specifications

Melting Point 178°C
Color White-Yellow
Quantity 1 g
UN Number 2811
Formula Weight 329.83
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Ondansetron Hydrochloride Dihydrate
RTECSNumber FE6375500
TSCA No
Recommended Storage Refrigerator