missing translation for 'onlineSavingsMsg'
Learn More

Ondansetron hydrochloride dihydrate, 98%

Catalog No. AC461160010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC461160010 1 g
AC461160050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC461160010 Supplier Thermo Scientific Chemicals Supplier No. 461160010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 103639-04-9 | HCl·2H2O | 365.85 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 103639-04-9
Molecular Formula HCl·2H2O
Molecular Weight (g/mol) 365.85
InChI Key BGGIFKYQGVGSIW-UHFFFAOYSA-N
Synonym 1,2,3,9-tetrahydro-9-methyl-3-2-methyl-1h-imidazol-1-yl methyl-4h-carbazol-4-one hydrochloride; gr 38032f; ondansetron hydrochloride
PubChem CID 71317260
IUPAC Name chlorine;9-methyl-3-[(2-methylimidazol-1-yl)methyl]-2,3-dihydro-1H-carbazol-4-one;dihydrate
SMILES CC1=NC=CN1CC2CCC3=C(C2=O)C4=CC=CC=C4N3C.O.O.[Cl]

Specifications

Melting Point 178.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 365.85
Percent Purity 98%
Chemical Name or Material Ondansetron hydrochloride dihydrate
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement Toxic if swallowed. Toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.

RUO – Research Use Only