missing translation for 'onlineSavingsMsg'
Learn More

OmniSolve Hexane, 99.8%, HR-GC Grade, MilliporeSigma™

Catalog No. MHX02971
Catalog No. MHX02971 Supplier Technidata Supplier No. US1HX0297/1

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 110-54-3
Molecular Formula C6H14
Molecular Weight (g/mol) 86.18
MDL Number MFCD02179311
InChI Key VLKZOEOYAKHREP-UHFFFAOYSA-N
Synonym n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, dipropyl, heksan, hexanen, hexanes
PubChem CID 8058
ChEBI CHEBI:29021
IUPAC Name hexane
SMILES CCCCCC

Specifications

Density 0.66kg/L
Physical Form Liquid
Assay Percent Range 99.80%
Linear Formula CH3(CH2)4CH3 with 2- and 3-methylpentanes and methylcyclopentane
Formula Weight 86.18
Grade HR-GC
Chemical Name or Material Hexane, 99.8%, HR-GC, OmniSolv™
Recommended Storage Room Temperature