missing translation for 'onlineSavingsMsg'
Learn More

OmniSolv™ Hexanes, ≤98.5%, MilliporeSigma™

Catalog No. MHX02961
Change view
Click to view available options
Quantity:
4 L
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MHX02961 4 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MHX02961 Supplier Technidata Supplier No. US1HX0296/1

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 110-54-3
Molecular Formula C6H14
Molecular Weight (g/mol) 86.18
MDL Number MFCD02179311
InChI Key VLKZOEOYAKHREP-UHFFFAOYSA-N
Synonym n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, hexanen, hexanes
PubChem CID 8058
ChEBI CHEBI:29021
IUPAC Name hexane
SMILES CCCCCC

Specifications

Density 0.66g/cm3 (20°C)
Quantity 4 L
Explosion Limit 1.0 to 8.1%(V)
Ignition Point 240°C
Packaging Glass Bottle
Linear Formula CH3(CH2)4CH3
Vapor Pressure 160 hPa (20°C)
Formula Weight 86.18
Chemical Name or Material Hexanes
Recommended Storage Store without restrictions