missing translation for 'onlineSavingsMsg'
Learn More

Diba Omnifit™ T-Series Bottle Caps

Catalog No. OF945T4F
Change view
Click to view available options
Thread Size (Metric):
GL 32
GL 38
GL 45
Includes:
2 Ports
2 Ports, Check Valve
2 Ports, No Valve
3 Ports
3 Ports, Check Valve
3 Ports, No Valve
4 Ports
4 Ports, Check Valve
4 Ports, No Valve
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Thread Size (Metric) Includes
OF945T4F GL 45 4 Ports, Check Valve
OF945T2 GL 45 2 Ports, No Valve
OF938T2F GL 38 2 Ports, Check Valve
OF945T3F GL 45 3 Ports, Check Valve
OF932T2 GL 32 2 Ports, No Valve
OF945T3 GL 45 3 Ports
OF938T4 GL 38 4 Ports, No Valve
OF945T2F GL 45 2 Ports, Check Valve
OF938T3 GL 38 3 Ports, No Valve
OF938T2 GL 38 2 Ports
OF945T4 GL 45 4 Ports
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Catalog No. OF945T4F Supplier Diba Supplier No. 00945T 4F
Only null left

Easy to use and versatile range of caps requires no fittings

  • Luer ported, straight through tubing
  • Simply push 1/8 in. O.D. hard wall tubing through the ports
  • For 1/16 in. O.D. hard wall tubing, use OF9LA adapter supplied with cap
  • Use OF9BA series adapter for softwall tubing
  • Anti-twist design prevents tubing from kinking when removing cap
  • Each port also accepts male Luer connectors
  • With polypropylene screw cap and chemically inert solid PTFE body

Not recommended for use under pressure with Luer products fitting as no Luer lock is employed.

Order Info

Available with Luer port or Luer port with fluorocarbon check valve and 10μm porosity PTFE inlet filter enabling air to flow into the bottle while preventing particulate contamination and preventing harmful vapors from escaping.

Specifications

Color Blue, White
Includes 4 Ports, Check Valve
Material PTFE
No. per Case 1 Cs.
Sterility Non-sterile
Thread Size (Metric) GL 45
Disposable Reusable
Height (Metric) 29.8 mm
Product Type Solvent Waste Cap
For Use With (Equipment) T series solvent bottle
Diameter (Metric) 54 mm
Autoclavable Not Autoclavable
Show More Show Less