Learn More
Omni International Omni Tip Hybrid Probe Replacement Shaft Clips
Description
- Replacement shaft clips
- Accessory
Specifications
Specifications
| Product Type | Replacement Shaft Clips |
| Autoclavable | Autoclavable |
| Material | PTFE |
| Description | Replacement Shaft Clips for the 12 mm Omni Tip Hybrid Probe |
| For Use With (Equipment) | 12 mm Omni Tip Hybrid Probe |
| Packaging Quantity | 25 Pk. |