missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Omics Clamp

Catalog No. 09927161
Change view
Click to view available options
For Use With (Equipment):
Tissue Disaggregator
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
09927161 Tissue Disaggregator
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09927161 Supplier Cytiva Supplier No. 29509355
NLSU: Get 1 Consumable Product When You Buy 1 Obtenez 1 produit consommable à l’achat d’un produit
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Provides a secure seal after sample insertion and supports the bag during tissue dissociation.

  • Provides a secure seal
  • Supports the bag during tissue dissociation

Specifications

Product Type Clamp
Autoclavable Not Autoclavable
For Use With (Equipment) Tissue Disaggregator

For research use only (RUO). Not for diagnostic use.