Learn More
Thermo Scientific™ Olympus™ 4X Objective, U Plan S-Apo, 0.16 NA
Catalog No. NX1004X
Encompass
Change view
Click to view available options
Quantity:
1 each
Catalog No. Quantity
NX1004X 1 each
1 options
Catalog No. NX1004X Supplier Thermo Scientific™ Supplier No. NX1004X
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Expand the functionality of your Thermo Scientific™ high-content instrument
Add a wide range of magnifications using Carl Zeiss™ and Olympus™ Objective Lenses for Thermo Scientific™ ArrayScan™ HCA Readers and the Thermo Scientific™ CellInsight™ NXT High Content Screening (HCS) Platform.
Specifications
Type Objective Lens
Product Line Olympus
Description Olympus™ 4X Objective, U Plan S-Apo (0.16 NA)
For Use With (Equipment) CellInsight CX5 HCS Platform
Lens Type Dry
Magnification 4x (0.16NA)
Quantity 1 each
For Research Use Only. Not for use in diagnostic procedures.