Learn More
Thermo Scientific™ Olympus™ 2X Objective, PlanApo, 0.08 NA
Numéro de catalogue. NX1002X
Encompass
Modifier l'affichage
Click to view available options
Quantity:
1 Ea.
Numéro de catalogue. Quantity
NX1002X 1 Ea.
1 options
Numéro de catalogue. NX1002X Fournisseur Thermo Scientific™ Code fournisseur NX1002X
Pour de laide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Expand the functionality of your Thermo Scientific™ high-content instrument
Add a wide range of magnifications using Carl Zeiss™ and Olympus™ Objective Lenses for Thermo Scientific™ ArrayScan™ HCA Readers and the Thermo Scientific™ CellInsight™ NXT High Content Screening (HCS) Platform.
The Olympus 2X (0.08NA) PlanApo objective lens is suitable for use with the Thermo Scientific CellInsight CX5, CX7 LED, and CX7 LZR high content screening (HCS) platforms. It has a magnification power suited for subcellular imaging and a numerical aperture that strikes a balance between resolution and versatility.
Spécifications
Type Objective Lens
Product Line Olympus
Description Olympus™ 2X Objective, PlanApo (0.08 NA)
For Use With (Equipment) CellInsight CX5 HCS Platform
Lens Type Dry
Magnification 2x (0.08NA)
Quantity 1 Ea.
For Research Use Only. Not for use in diagnostic procedures.