missing translation for 'onlineSavingsMsg'
Learn More

Olopatadine Hydrochloride 98.0+%, TCI America™

Catalog No. O03611G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O03611G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O03611G Supplier TCI America Supplier No. O03611G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 140462-76-6
Molecular Formula C21H24ClNO3
Molecular Weight (g/mol) 373.877
MDL Number MFCD00875716
InChI Key HVRLZEKDTUEKQH-NOILCQHBSA-N
Synonym olopatadine hydrochloride
PubChem CID 5282402
IUPAC Name 2-[(11Z)-11-[3-(dimethylamino)propylidene]-6H-benzo[c][1]benzoxepin-2-yl]acetic acid;hydrochloride
SMILES CN(C)CCC=C1C2=CC=CC=C2COC3=C1C=C(C=C3)CC(=O)O.Cl

Specifications

Melting Point 248°C
Color White-Yellow
Quantity 1 g
Formula Weight 373.88
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Olopatadine Hydrochloride
RTECSNumber HQ4135060
TSCA No