missing translation for 'onlineSavingsMsg'
Learn More

Olive oil, pure, virgin

Catalog No. AC416540250
Change view
Click to view available options
Quantity:
25 mL
500 mL
2.5 L
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC416540250 25 mL Glass bottle
AC416545000 500 mL Glass bottle
AC416540025 2.5 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC416540250 Supplier Thermo Scientific Chemicals Supplier No. 416540250

CAS: 8001-25-0

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 8001-25-0

Specifications

Acidity 2.0mg KOH/g max.
Absorbance 0.2 (270nm, 1g/100ml cyclohexane) max. at 425nm
Color Yellow
Density 0.9135g/mL
Flash Point 225°C
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4670 to 1.4710
Quantity 25 mL
Saponification Value 184 to 196mg KOH/g
Merck Index 14,6836
Specific Gravity 0.9135
Solubility Information Solubility in water: .. Other solubilities: practically insoluble in alcohol, miscible with light petroleum (50-60°C)
Grade Pure
Concentration or Composition (by Analyte or Components) of fatty acids: 0.1 % max. (< C16) 7.5 to 20.0 % (Palmitic acid) 3.5 % max. (Palmitoleic acid) 0.5 to 5.0 % (Stearic acid) 56.0 to 85.0 % (Oleic acid) 3.5 to 20.0 % (Linoleic acid) 1.2 % max. (Linolenic acid) 0.7 % max. (Arachidic acid) 0.4 % max. (Eic
Physical Form Powder
Chemical Name or Material Olive oil
Show More Show Less
EINECSNumber 232-277-

RUO – Research Use Only