missing translation for 'onlineSavingsMsg'
Learn More

Olive Oil, NF, Spectrum™ Chemical

Catalog No. 18604885
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
20 L
200 L
4 L
Packaging:
Amber Glass Bottle
Drum
Poly Drum
Catalog No. Quantity Packaging
18604885 20 L Poly Drum
18604884 4 L Amber Glass Bottle
18604886 200 L Drum
3 options

Catalog No. 18604885

Supplier: Spectrum Chemical Mfg Cor OL13020LTPL

Only null left

OL130, 8001-25-0

Spectrum™ Chemical Olive Oil, NF is used as a vehicle for oily injection suspensions and as an emollient. Spectrum Chemical manufactured NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 8001-25-0

Specifications

CAS Max % 1
Packaging Poly Drum
Quantity 20 L
Grade NF