missing translation for 'onlineSavingsMsg'
Learn More

Olive Oil, NF, Spectrum™ Chemical

Catalog No. 18604884
Change view
Click to view available options
Quantity:
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
Drum
Poly Drum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18604884 4 L Amber Glass Bottle
18604885 20 L Poly Drum
18604886 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18604884 Supplier Spectrum Chemical Mfg Cor Supplier No. OL1304LTGL

OL130, 8001-25-0

Spectrum™ Chemical Olive Oil, NF is used as a vehicle for oily injection suspensions and as an emollient. Spectrum Chemical manufactured NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 8001-25-0

Specifications

CAS Max % 1
Packaging Amber Glass Bottle
Quantity 4 L
Grade NF