missing translation for 'onlineSavingsMsg'
Learn More

Olive Oil, NF, Spectrum™ Chemical

Catalog No. 18604882
Change view
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18604882 500 mL Amber Glass Bottle
18604883 1 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18604882 Supplier Spectrum Chemical Mfg Cor Supplier No. OL130500MLGL

OL130, 8001-25-0

Spectrum™ Chemical Olive Oil, NF is used as a vehicle for oily injection suspensions and as an emollient. Spectrum Chemical manufactured NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 8001-25-0

Specifications

CAS Max % 1
Packaging Amber Glass Bottle
Quantity 500 mL
Grade NF