missing translation for 'onlineSavingsMsg'
Learn More

Oleylamine 50.0+%, TCI America™

Catalog No. p-7134163
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O005925ML 25 mL
O0059500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O005925ML Supplier TCI America Supplier No. O005925ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 112-90-3
Molecular Formula C18H37N
Molecular Weight (g/mol) 267.50
MDL Number MFCD00066507
InChI Key QGLWBTPVKHMVHM-KTKRTIGZSA-N
Synonym 9-octadecenylamine, 9-octadecen-1-amine, cis-9-octadecenylamine, e-octadec-9-en-1-amine, 1-amino-9-octadecene, 9e octadec-9-enylamine, 9-octadecenylamine 8ci, 36505-83-6 hydrofluoride, 3811-68-5 unspecified acetate
PubChem CID 6258392
IUPAC Name (9Z)-octadec-9-en-1-amine
SMILES CCCCCCCC\C=C/CCCCCCCCN

Specifications

Color Yellow
Quantity 25 mL
UN Number 2735
Formula Weight 267.50
Percent Purity ≥50.0% (GC)
Physical Form Liquid
Chemical Name or Material Oleylamine
EINECSNumber (2)-0133&(2)-0185&(2)-0176
RTECSNumber RG4130000
TSCA Yes