missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich Oleylamine

Catalog No. 9098315G
Encompass
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
9098315G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 9098315G Supplier Sigma Aldrich Supplier No. 9098315G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Oleylamine

Specifications

CAS 112-90-3
Melting Point 18°C to 26°C
Density 0.813 g/mL (at 25°C)
Boiling Point 348°C to 350°C
Molecular Formula C18H37N
Refractive Index n20/D 1.4596 (literature)
Linear Formula CH3(CH2)7 CH=CH(CH2)7 CH2NH2
MDL Number MFCD00066507
Quantity 5 g
Synonym cis-1-Amino-9-octadecene
Molecular Weight (g/mol) 267.49
Show More Show Less