missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Oleyl Chloride 70.0+%, TCI America™
Specifications
Specifications
| CAS | 16507-61-2 |
| Color | Yellow |
| Molecular Formula | C18H35Cl |
| Quantity | 25 mL |
| MDL Number | MFCD00058939 |
| InChI Key | IFABLCIRROMTAN-MDZDMXLPSA-N |
| SMILES | CCCCCCCCC=CCCCCCCCCCl |
| IUPAC Name | (E)-1-chlorooctadec-9-ene |
| Molecular Weight (g/mol) | 286.928 |
| PubChem CID | 6536917 |
| Show More |
Safety and Handling
| TSCA | No |