missing translation for 'onlineSavingsMsg'
Learn More

Oleyl Alcohol 60.0+%, TCI America™

Catalog No. p-7134162
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O005825ML 25 mL
O0058500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O005825ML Supplier TCI America Supplier No. O005825ML
Only null left

Chemical Identifiers

CAS 143-28-2
Molecular Formula C18H36O
Molecular Weight (g/mol) 268.49
MDL Number MFCD00002993
InChI Key ALSTYHKOOCGGFT-KTKRTIGZSA-N
Synonym oleyl alcohol, cis-9-octadecen-1-ol, z-octadec-9-en-1-ol, ocenol, conditioner 1, dermaffine, lancol, novol, oceol, oleol
PubChem CID 5284499
ChEBI CHEBI:73504
IUPAC Name (9Z)-octadec-9-en-1-ol
SMILES CCCCCCCC\C=C/CCCCCCCCO

Specifications

Color Yellow
Boiling Point 207°C
Quantity 25 mL
Formula Weight 268.49
Percent Purity ≥60.0% (GC)
Physical Form Liquid
Chemical Name or Material Oleyl Alcohol
EINECSNumber (2)-0258
RTECSNumber RG4120000
TSCA Yes