missing translation for 'onlineSavingsMsg'
Learn More

Oleyl alcohol, ca. 60%, technical

Catalog No. p-3758607
Change view
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC129530010 1 L Glass Bottle
AC129530025 2.5 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC129530010 Supplier Thermo Scientific Chemicals Supplier No. 129530010
Only null left

CAS: 143-28-2 | C18H36O | 268.49 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 143-28-2
Molecular Formula C18H36O
Molecular Weight (g/mol) 268.49
MDL Number MFCD00002993
InChI Key ALSTYHKOOCGGFT-KTKRTIGZSA-N
Synonym oleyl alcohol, cis-9-octadecen-1-ol, z-octadec-9-en-1-ol, ocenol, conditioner 1, dermaffine, lancol, novol, oceol, oleol
PubChem CID 5284499
ChEBI CHEBI:73504
IUPAC Name (9Z)-octadec-9-en-1-ol
SMILES CCCCCCCC\C=C/CCCCCCCCO

Specifications

Melting Point 5.0°C
Color Colorless to Yellow
Density 0.8400g/mL
Boiling Point 207.0°C (13.0 mmHg)
Flash Point 170°C
Infrared Spectrum Authentic
Assay Percent Range 60% Typical (GC)
Packaging Glass Bottle
Linear Formula CH3(CH2)7CH=CH(CH2)8OH
Refractive Index 1.4590 to 1.4640
Quantity 1 L
Beilstein 01, 453
Specific Gravity 0.84
Merck Index 15, 6924
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in alcohol and ether, soluble in warbontetrachloride
Formula Weight 268.47
Percent Purity ca. 60%
Grade Technical
Physical Form Viscous Liquid
Chemical Name or Material Oleyl alcohol
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 205-597-3

RUO – Research Use Only