missing translation for 'onlineSavingsMsg'
Learn More

Oleyl alcohol, 99+%

Catalog No. p-7032125
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3347503 1 g
AAH3347506 5 g
AAH3347514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 missing translation for 'options'
Catalog No. AAH3347503 missing translation for 'mfr' Thermo Scientific Chemicals missing translation for 'supplierNo' H3347503

CAS: 143-28-2 | C18H36O | 268.49 g/mol

Oleyl alcohol is used in softening and lubrication of textile fabrics, in the production of carbon paper, stencil paper, and printing ink. It finds application in cosmetic products viz skin creams, lotions as a thickner, and in hair conditioners and hair coating shampoos. It is utilized as an antifoaming agent and cutting lubricant, as the precursor for the preparation of its sulfuric ester derivatives, which are used in detergents and wetting agents. It plays a vital role in various formulations for drug delivery.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Oleyl alcohol is used in softening and lubrication of textile fabrics, in the production of carbon paper, stencil paper, and printing ink. It finds application in cosmetic products viz skin creams, lotions as a thickner, and in hair conditioners and hair coating shampoos. It is utilized as an antifoaming agent and cutting lubricant, as the precursor for the preparation of its sulfuric ester derivatives, which are used in detergents and wetting agents. It plays a vital role in various formulations for drug delivery.

Solubility
Miscible with alcohol and ether. Slightly miscible with carbon tetrachloride. Immiscible with water.

Notes
Incompatible with oxidizing agents and strong acids.

Chemical Identifiers

CAS 143-28-2
Molecular Formula C18H36O
Molecular Weight (g/mol) 268.49
MDL Number MFCD00002993
InChI Key ALSTYHKOOCGGFT-KTKRTIGZSA-N
Synonym oleyl alcohol, cis-9-octadecen-1-ol, z-octadec-9-en-1-ol, ocenol, conditioner 1, dermaffine, lancol, novol, oceol, oleol
PubChem CID 5284499
ChEBI CHEBI:73504
IUPAC Name (Z)-octadec-9-en-1-ol
SMILES CCCCCCCC\C=C/CCCCCCCCO

Specifications

Melting Point ∼6°C
Density 0.849
Boiling Point 206°C to 207°C (13 mmHg)
Flash Point 180°C (356°F)
Refractive Index 1.46
Quantity 1 g
Beilstein 1723962
Solubility Information Miscible with alcohol and ether. Slightly miscible with carbon tetrachloride. Immiscible with water.
Formula Weight 268.48
Percent Purity ≥99%
Chemical Name or Material Oleyl alcohol
Show More Show Less
missing translation for 'healthHazard1' H315-H319
missing translation for 'healthHazard2' GHS H Statement
H315
Causes skin irritation.
missing translation for 'healthHazard3' P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
EINECSNumber 205-597-3
RTECSNumber RG4120000
TSCA Yes
Recommended Storage Store at -20°C

RUO – Research Use Only