missing translation for 'onlineSavingsMsg'
Learn More

Oleoyl Chloride 80.0+%, TCI America™

Catalog No. p-7134157
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O005325G 25 g
O0053250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O005325G Supplier TCI America Supplier No. O005325G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 112-77-6
Molecular Formula C18H33ClO
Molecular Weight (g/mol) 300.91
MDL Number MFCD00134332
InChI Key MLQBTMWHIOYKKC-KTKRTIGZSA-N
Synonym elaidoylchloride, elaidoyl chloride, 9-octadecenoyl chloride, oleoylchloride, elaidic acid chloride, trans-9-octadecenoyl chloride, e-octadec-9-enoyl chloride, 9e-octadec-9-enoyl chloride
PubChem CID 5702750
IUPAC Name (9Z)-octadec-9-enoyl chloride
SMILES CCCCCCCC\C=C/CCCCCCCC(Cl)=O

Specifications

Color Yellow
Boiling Point 200°C
Quantity 25 g
UN Number 3265
Formula Weight 300.91
Percent Purity ≥80.0% (GC)
Physical Form Liquid
Chemical Name or Material Oleoyl Chloride
EINECSNumber (2)-3552
TSCA Yes
Recommended Storage Refrigerator