missing translation for 'onlineSavingsMsg'
Learn More

Oleamide 65.0+%, TCI America™

Catalog No. O010725G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O010725G 25 g
O0107500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O010725G Supplier TCI America Supplier No. O010725G

Chemical Identifiers

CAS 301-02-0
Molecular Formula C18H35NO
Molecular Weight (g/mol) 281.484
MDL Number MFCD00053638
InChI Key FATBGEAMYMYZAF-KTKRTIGZSA-N
Synonym oleamide, oleic acid amide, oleylamide, oleyl amide, adogen 73, crodamide o, slip-eze, armoslip cp, crodamide or, cis-9,10-octadecenoamide
PubChem CID 5283387
ChEBI CHEBI:116314
IUPAC Name (Z)-octadec-9-enamide
SMILES CCCCCCCCC=CCCCCCCCC(=O)N

Specifications

Melting Point 70°C
Color White-Yellow
Quantity 25 g
Formula Weight 281.48
Percent Purity ≥65.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Oleamide
EINECSNumber (2)-0824&(2)-0976
RTECSNumber RG2230000
TSCA Yes