missing translation for 'onlineSavingsMsg'
Learn More

Olanzapine 98.0+%, TCI America™

Catalog No. O03931G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O03931G 1 g
O03935G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O03931G Supplier TCI America Supplier No. O03931G

Chemical Identifiers

CAS 132539-06-1
Molecular Formula C17H20N4S
Molecular Weight (g/mol) 312.44
MDL Number MFCD00866702
InChI Key WXPNDRBBWZMPQG-UHFFFAOYSA-N
Synonym olanzapine, zyprexa, zyprexa zydis, olansek, zalasta, symbyax, zyprexa intramuscular, zyprexa velotab, 2-methyl-4-4-methylpiperazin-1-yl-10h-benzo b thieno 2,3-e 1,4 diazepine, olanzapina
PubChem CID 4585
IUPAC Name 5-methyl-8-(4-methylpiperazin-1-yl)-4-thia-2,9-diazatricyclo[8.4.0.0³,⁷]tetradeca-1(14),2,5,7,10,12-hexaene
SMILES CN1CCN(CC1)C1=C2C=C(C)SC2=NC2=CC=CC=C2N1

Specifications

Melting Point 194°C
Color Yellow
Quantity 1 g
UN Number 2811
Formula Weight 312.44
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Olanzapine
RTECSNumber XJ9007750
TSCA No
Recommended Storage Refrigerator