missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Olanzapine

Chemical Identifiers

CAS 132539-06-1
Molecular Formula C17H20N4S
Molecular Weight (g/mol) 312.44
MDL Number MFCD00866702
InChI Key WXPNDRBBWZMPQG-UHFFFAOYSA-N
IUPAC Name 5-methyl-8-(4-methylpiperazin-1-yl)-4-thia-2,9-diazatricyclo[8.4.0.0³,⁷]tetradeca-1(14),2,5,7,10,12-hexaene
SMILES CN1CCN(CC1)C1=C2C=C(C)SC2=NC2=CC=CC=C2N1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC464290010
View Documents
Thermo Scientific™
464290010
1 g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC464290050
View Documents
Thermo Scientific™
464290050
5 g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

132539-06-1
312.44
WXPNDRBBWZMPQG-UHFFFAOYSA-N
CN1CCN(CC1)C1=C2C=C(C)SC2=NC2=CC=CC=C2N1
C17H20N4S
MFCD00866702
5-methyl-8-(4-methylpiperazin-1-yl)-4-thia-2,9-diazatricyclo[8.4.0.0³,⁷]tetradeca-1(14),2,5,7,10,12-hexaene

Specifications

132539-06-1
MFCD00866702
Solubility aq. soln.: insoluble. Other solubilities: soluble in hot acetone
CN1CCN(CC1)C1=C2C=C(C)SC2=NC2=CC=CC=C2N1
312.44
Olanzapine
C17H20N4S
1 g
WXPNDRBBWZMPQG-UHFFFAOYSA-N
5-methyl-8-(4-methylpiperazin-1-yl)-4-thia-2,9-diazatricyclo[8.4.0.0³,⁷]tetradeca-1(14),2,5,7,10,12-hexaene
312.43
Videos
Safety and Handling

Safety and Handling

GHS H Statement Toxic if swallowed.

Causes skin irritation.

Causes serious eye irritation.

GHS P Statement IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.

IF ON SKIN: Wash with plenty of soap and water.

If skin irritation occurs: Get medical advice/attention.

If eye irritation persists: Get medical advice/attention.

Wear protective gloves/protective clothing/eye protection/face protection.

GHS Signal Word: Danger

SDS
Documents

Documents

RUO – Research Use Only