missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Olanzapine

Catalog No. AC464290010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC464290010 1 g
AC464290050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC464290010 Supplier Thermo Scientific Chemicals Supplier No. 464290010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 132539-06-1 | C17H20N4S | 312.44 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 132539-06-1
Molecular Formula C17H20N4S
Molecular Weight (g/mol) 312.44
MDL Number MFCD00866702
InChI Key WXPNDRBBWZMPQG-UHFFFAOYSA-N
IUPAC Name 5-methyl-8-(4-methylpiperazin-1-yl)-4-thia-2,9-diazatricyclo[8.4.0.0³,⁷]tetradeca-1(14),2,5,7,10,12-hexaene
SMILES CN1CCN(CC1)C1=C2C=C(C)SC2=NC2=CC=CC=C2N1

Specifications

Melting Point 194.0°C
Quantity 1 g
Solubility Information Solubility aq. soln.: insoluble. Other solubilities: soluble in hot acetone
Formula Weight 312.43
Chemical Name or Material Olanzapine
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement Toxic if swallowed.

Causes skin irritation.

Causes serious eye irritation.

Precautionary Statement GHS P Statement IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.

IF ON SKIN: Wash with plenty of soap and water.

If skin irritation occurs: Get medical advice/attention.

If eye irritation persists: Get medical advice/attention.

Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only