missing translation for 'onlineSavingsMsg'
Learn More

Oil Red O BioReagent, Suitable for Electrophoresis, MilliporeSigma™ Supelco™

Catalog No. 111012938
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012938 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012938 Supplier Merck Emd Millipore Supplier No. O975525G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

MDL Number MFCD00003898
Synonym 1-([4-(Xylylazo)xylyl]azo)-2-naphthol; 1-[2,5-Dimethyl-4-(2,5-dimethylphenylazo)phenylazo]-2-naphthol; Solvent Red 27; Solvent Red 27

Specifications

Melting Point 120°C
CAS 1320-06-5
Packaging Glass Bottle
Linear Formula C26H24N4 O
Quantity 25 g
Formula Weight 408.49