missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Oil Red O

Catalog No. p-7020882
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1298914 25 g
AAA1298922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1298914 Supplier Thermo Scientific Chemicals Supplier No. A1298914

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1320-06-5 | C26H24N4O | 408.51 g/mol

Oil Red O is suitable as a lipid and lipoprotein stain on cellulose acetate. It is one of the diazo dyes used for staining of neutral triglycerides and lipids. It is used to analyze muscle fibers. It acts as an agent that stains lipids, muscle fibers and alveolar macrophages. Further, it is also used as marking dye. It is an active component of red colored smokes in pyrotechnics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Oil Red O is suitable as a lipid and lipoprotein stain on cellulose acetate. It is one of the diazo dyes used for staining of neutral triglycerides and lipids. It is used to analyze muscle fibers. It acts as an agent that stains lipids, muscle fibers and alveolar macrophages. Further, it is also used as marking dye. It is an active component of red colored smokes in pyrotechnics.

Solubility
Soluble in ethanol:chloroform (1:1). Insoluble in water.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 1320-06-5
Molecular Formula C26H24N4O
Molecular Weight (g/mol) 408.51
MDL Number MFCD00003898
InChI Key HJAYEODIXUYVIC-LTFAAIOASA-N
Synonym C.I. 26125; Solvent Red 27
PubChem CID 6046885
IUPAC Name (1Z)-1-[[4-[(2,5-dimethylphenyl)diazenyl]-2,5-dimethylphenyl]hydrazinylidene]naphthalen-2-one
SMILES CC1=CC=C(C)C(=C1)N=NC1=CC(C)=C(N\N=C2/C(=O)C=CC3=CC=CC=C23)C=C1C

Specifications

Melting Point ∼166°C (decomposition)
Beilstein 3491382
Solubility Information Soluble in ethanol:chloroform (1:1). Slightly soluble in water.
Quantity 25 g
Formula Weight 408.51
Chemical Name or Material Oil Red O
EINECSNumber 215-295-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only