missing translation for 'onlineSavingsMsg'
Learn More

Oil Orange SS, TCI America™

Catalog No. T055325G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T055325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T055325G Supplier TCI America Supplier No. T055325G
Only null left

Chemical Identifiers

CAS 2646-17-5
Molecular Formula C17H14N2O
Molecular Weight (g/mol) 262.312
MDL Number MFCD00059523
InChI Key YARMASSTSUTDQJ-ZPHPHTNESA-N
Synonym Solvent Orange 2, 1-(o-Tolylazo)-2-naphthol
PubChem CID 5835026
IUPAC Name (1Z)-1-[(2-methylphenyl)hydrazinylidene]naphthalen-2-one
SMILES CC1=CC=CC=C1NN=C2C(=O)C=CC3=CC=CC=C32

Specifications

Melting Point 128°C
Quantity 25 g
Formula Weight 262.31
Physical Form Crystalline Powder
Chemical Name or Material Oil Orange SS
EINECSNumber (5)-3065
RTECSNumber QL5425000
TSCA Yes