missing translation for 'onlineSavingsMsg'
Learn More

Oil, for heat transfer

Catalog No. AC268770025
Change view
Click to view available options
Quantity:
2.5 L
10 L
Packaging:
Glass bottle
Plastic drum
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC268770025 2.5 L Glass bottle
AC268770100 10 L Plastic drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC268770025 Supplier Thermo Scientific Chemicals Supplier No. 268770025

CAS: 64742-65- | C16H10N2Na2O7S2 | 452.363 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 64742-65-
Molecular Formula C16H10N2Na2O7S2
Molecular Weight (g/mol) 452.363
MDL Number MFCD00147787
InChI Key AEOVEGJBKQQFOP-DDVLFWKVSA-L
Synonym acid orange 10, wool orange 2g, orange g, c.i. acid orange 10, c.i. orange g, c.i. food orange 4, light orange g, colacid orange g, dolkwal orange g, hexacol orange g
PubChem CID 9566064
IUPAC Name disodium;(8Z)-7-oxo-8-(phenylhydrazinylidene)naphthalene-1,3-disulfonate
SMILES C1=CC=C(C=C1)NN=C2C(=O)C=CC3=CC(=CC(=C32)S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+]

Specifications

Melting Point -6.0°C
Color Yellow
Density 0.8600g/mL
Flash Point 230°C
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 2.5 L
Specific Gravity 0.86
Solubility Information Solubility in water: insoluble.
Viscosity 30 mm2/s (40°C)
Physical Form Viscous Liquid
Chemical Name or Material Oil
Show More Show Less
EINECSNumber 265-169-7

RUO – Research Use Only