missing translation for 'onlineSavingsMsg'
Learn More

Ofloxacin

Catalog No. AAJ6208003
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6208003 1 g
AAJ6208006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6208003 Supplier Thermo Scientific Chemicals Supplier No. J6208003

Fluorinated nalidixic acid analog with broad-spectrum antibacterial activity | CAS: 82419-36-1 | C18H20FN3O4 | 361.373 g/mol

Fluorinated nalidixic acid analog with broad-spectrum antibacterial activity

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 82419-36-1
Molecular Formula C18H20FN3O4
Molecular Weight (g/mol) 361.373
MDL Number MFCD00226105
InChI Key GSDSWSVVBLHKDQ-UHFFFAOYSA-N
Synonym Oflocet; Ofloxacine
PubChem CID 4583
ChEBI CHEBI:7731
SMILES CC1COC2=C3N1C=C(C(=O)C3=CC(=C2N4CCN(CC4)C)F)C(=O)O

Specifications

Type Ofloxacin
Physical Form Crystalline Powder
Melting Point 270°C to 275°C
Color White to Yellow
Quantity 1 g
Merck Index 14,6771
Solubility Information Soluble in acetic acid or water; Slightly soluble in methanol
Formula Weight 361.37
Chemical Name or Material Ofloxacin
RTECSNumber UU8815500
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only